Sensidyne 126SH Gas Detection Tubes, Carbon dioxide
Representative image only.
Sensidyne 126SH Gas Detection Tubes, Carbon dioxide
Sensidyne | Mfr: 126SH| Item: 8190015
Measure a wide variety of gases and vapors with this detection system
$139.97CAD / Box of 10
$139.97CAD / Box of 10
Buy It Together
Current Item
Sensidyne 126SH Gas Detection Tubes, Carbon dioxide
Sensidyne 126SH Gas Detection Tubes, Carbon dioxide
8190015
$139.97CAD / Box of 10
+
Cannon 9721-B56 Viscometer, universal size 75
9893452
$329.23CAD / Each
=
Total Price 469.20 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Hundreds of direct-read tubes available to meet your changing needs
  • 2 year warranty
More About this Item
Stable absorbing media ensure distinct color changes for clear, accurate results. Calibration scales are printed on the basis of individual production lot, eliminating variation in tube diameter, packing precision, and reactivity.
Two year warranty
Specifications
  • Gas MeasuredCarbon dioxide
  • Range1 to 20%
  • TLV (ppm)5000
  • DescriptionGas Detection Tubes, Carbon dioxide