Sellstrom Faceshield with Headgear, Clear PC Shield
Representative image only.
Sellstrom Faceshield with Headgear, Clear PC Shield
Sellstrom | Mfr: S32010| Item: 8647800
Compact shield provides protection without restricting movement
$98.72CAD / Each
BUY MORE & SAVE
  • 1-9$98.72 CAD / Each
  • 10+$88.85 CAD / Each
$98.72CAD / Each
BUY MORE & SAVE
  • 1-9$98.72 CAD / Each
  • 10+$88.85 CAD / Each
Buy It Together
Current Item
Sellstrom Faceshield with Headgear, Clear PC Shield
Sellstrom Faceshield with Headgear, Clear PC Shield
8647800
$98.72CAD / Each
+ =
Total Price 146.75 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Face shield curves naturally to fit facial contours ensuring reliable protection from splash, particles, and flying debris
  • Headgear features a ratchet suspension which adjusts the size with the turn of a knob
  • Soft padded brow guard rests comfortably on the face
  • Polycarbonate shield is impact and splash resistant
  • Filters 99.9% of harmful UV
  • Headgear comes fully assembled
  • Compliance: ANSI Z87.1-2003, cUL certified to meet CAN/CSA Z94.3 standard
  • 1 year warranty
More About this Item
Faceshield curves naturally to fit facial contours — ensuring reliable protection from splash, particles, and flying debris. Headgear includes ratchet suspension to adjust size and tighten at the turn of a knob.
Soft, padded browguard rests comfortably on face. Rugged, polycarbonate shield is impact- and splash-resistant. Item comes fully assembled with headgear and shield included. Replacement shields are sold separately.
Compliance: ANSI Z87.1
1 Year Warranty
Specifications
  • Frame ColorClear
  • Lens ColorClear
  • ComplianceANSI Z87.1
  • DescriptionFaceshield with Headgear, Clear PC Shield