Sellstrom 31140 Laboratory faceshield, double crown w/acetate shield
Representative image only.
Sellstrom 31140 Laboratory faceshield, double crown w/acetate shield
Sellstrom | Mfr: S31140| Item: 8647822
Ultimate in lightweight face protection
$160.97CAD / Each
BUY MORE & SAVE
  • 1-9$160.97 CAD / Each
  • 10+$144.87 CAD / Each
$160.97CAD / Each
BUY MORE & SAVE
  • 1-9$160.97 CAD / Each
  • 10+$144.87 CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
White ABS plastic crown includes ratchet headgear and deluxe perspiration band to maintain a cool, comfortable fit. Lightweight clear acetate window (included) is fog-free to allow maximum visibility. Ideal for laboratory use. Select single-crown faceshield for protection from splash and particles. Select double-crown faceshield with chin protector for added impact protection.
1 Year Warranty
Specifications
  • Frame ColorWhite
  • Lens ColorClear
  • DescriptionLaboratory faceshield, double crown w/acetate shield