Self-Closing Check Valve, PVC, 1/2" NPT(F)

Representative image only.

Self-Closing Check Valve, PVC, 1/2" NPT(F)

| Mfr: CKM050V-PV| Item: 4852716

Patented diaphragm design does not depend on gravity, mounting position, or reverse flow to seal

$212.17CAD / Each

$212.17CAD / Each
Product Description

Key Features

  • Leak-free sealing prevents reverse flow
  • Create a secure seal using pipe thread connections
  • Diaphragm automatically positions itself against seat in the identical location
  • Diaphragm begins to open at approximately 1 to 1.5 psi
  • Flushing action of the inlet flow keeps the sealing area clean
  • 1/2" NPT(F) x 1/2" NPT(F) connections
  • PVC body
  • 1 year warranty

More About this Item

Use the self-closing diaphragm check valves to protect against hazardous and corrosive reverse flow. The diaphragm reseats against the seal without relying on gravity, mounting position, or reverse flow, ensuring reliable sealing even at low pressures. A union nut allows easy inspection and removal with minimal piping disruption. No loose or sliding internal parts eliminate chattering and help keep operation quiet. As inlet flow passes through the valve, both the diaphragm and valve seat are kept clean by the flushing action, keeping the sealing area clean and free of particles, minimizing the risk of leakage.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

1 Year Warranty
Specifications
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(F) x NPT(F)
  • Inlet Port1/2" NPT(F)
  • Seal MaterialViton®
  • Housing MaterialPVC
  • Max Pressure (PSI)150
  • Max Pressure (bar)9.6
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC body and Viton® disc
  • Flow PatternStraight
  • Length (in)3.8125
  • Diameter (in)2.375
  • DescriptionSelf-Closing Check Valve, PVC, 1/2" NPT(F)
  • Warranty1 year
Reviews
Q&A