SciMatCo SC9042 Storage Cabinet, Acid, Mini 20 L

Representative image only.

SciMatCo SC9042 Storage Cabinet, Acid, Mini 20 L

SciMatCo | Mfr: SC9042| Item: 8643400

Compact size is perfect for laboratory benchtop chemical storage

$1,841.74CAD / Each

$1,841.74CAD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Rugged cabinets are constructed of 1” (2.5 cm) thick, high-density plywood and finished with clear epoxy paint for superior durability. Liquid-tight, 2” (5.1 cm) trough at base of cabinet contains any spillage. Color and highly visible 8” (20.3 cm) hazard label ensures easy identification of cabinet type. These versatile cabinets are designed for the laboratory bench but also can be stacked three high, using optional floor stand, for a complete chemical storage center.

1 Year Warranty
Specifications
  • MaterialPlywood
  • Capacity (Liters)20
  • Capacity (Gal)5.3
  • Width (in)31
  • Height (in)17
  • Depth (InchShort)14 1/2
  • Width (cm)78.7
  • Height (cm)43.2
  • Depth (cm)36.8
  • ColorBlue
  • ComplianceOSHA, NFPA (flammable cabinet), UFC, IFC
  • Shipping Weight (lbs)60
  • DescriptionStorage Cabinet, Acid, Mini
  • Warranty1 Year
Reviews
Q&A