SciMatCo SC8079 Flammable Storage Cabinet, 24 gallon capacity, self-closing door

Representative image only.

SciMatCo SC8079 Flammable Storage Cabinet, 24 gallon capacity, self-closing door

SciMatCo | Mfr: SC8079| Item: 8178027

Wooden cabinets provide excellent thermal insulation in case of fire

$2,151.15CAD / Each

$2,151.15CAD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Use this lockable, wooden cabinet to conveniently store flammable liquids. Constructed of 1" (2.5 cm) thick exterior grade plywood; finished with yellow epoxy paint inside and out. Specially designed floor serves as a liquid-tight trough to retain spills. Locking door features continuous piano-type hinges, and a 8" x 8" (20.3 x 20.3 cm) "Flammable" hazard label. Contains one variable position shelf. Meets all applicable OSHA and NFPA standards for flammable safety cabinets. UL Listed.

Meet all applicable OSHA and NFPA standards for flammable safety cabinets. Models with self-closing door also meet the Uniform Fire Code (UFC). UL Listed.

Compliance: OSHA, NFPA, UFC

Approvals: UL Listed

1 Year Warranty
Specifications
  • MaterialWood
  • Capacity (Liters)90.8
  • Capacity (Gal)24
  • Number of Shelves1
  • Width (in)30
  • Height (in)32
  • Depth (InchShort)18 1/2
  • Width (cm)76.2
  • Height (cm)81.3
  • Depth (cm)47
  • ColorYellow
  • Self-ClosingYes
  • ComplianceOSHA, NFPA, UFC, UL Listed
  • Shipping Weight (lbs)120
  • DescriptionFlammable Storage Cabinet, 24 gallon capacity, self-closing door
  • Warranty1 Year
Reviews
Q&A