SciMatCo SC5060 Polypropylene Acid Storage Cabinet, 100 X 2.5 L bottle capacity

Representative image only.

SciMatCo SC5060 Polypropylene Acid Storage Cabinet, 100 X 2.5 L bottle capacity

SciMatCo | Mfr: SC5060| Item: 8178042

Liquid tight to prevent leakage

$6,180.87CAD / Each

$6,180.87CAD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

These polypropylene acid storage cabinets contain no metal parts— they won’t corrode or rust. Constructed of 3/8" (0.96 cm)-thick white polypropylene, cabinets feature plastic hinges and welded interior seams to prevent leakage. All cabinets include hazard warning label.

Compliance: OSHA, UFC, IFC

1 Year Warranty
Specifications
  • MaterialPolypropylene
  • Capacity (Liters)250
  • Capacity (Gal)66
  • Number of Shelves3
  • Width (in)48
  • Height (in)60
  • Depth (InchShort)18
  • Width (cm)121.9
  • Height (cm)152.4
  • Depth (cm)45.7
  • ColorWhite
  • ComplianceOSHA, UFC, IFC
  • Shipping Weight (lbs)210
  • DescriptionPolypropylene Acid Storage Cabinet, 100 X 2.5 L bottle capacity
  • Warranty1 Year
Reviews
Q&A