SciMatCo SC5000 Polypropylene Acid Storage Cabinet, 6 X 2.5 L bottle capacity

Representative image only.

SciMatCo SC5000 Polypropylene Acid Storage Cabinet, 6 X 2.5 L bottle capacity

SciMatCo | Mfr: SC5000| Item: 8178015

Liquid tight to prevent leakage

$1,028.43CAD / Each

$1,028.43CAD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

These polypropylene acid storage cabinets contain no metal parts— they won’t corrode or rust. Constructed of 3/8" (0.96 cm)-thick white polypropylene, cabinets feature plastic hinges and welded interior seams to prevent leakage. All cabinets include hazard warning label.

Compliance: OSHA, UFC, IFC

1 Year Warranty
Specifications
  • MaterialPolypropylene
  • Capacity (Liters)15
  • Capacity (Gal)4
  • Number of Shelves0
  • Width (in)16
  • Height (in)16
  • Depth (InchShort)16
  • Width (cm)40.6
  • Height (cm)40.6
  • Depth (cm)40.6
  • ColorWhite
  • ComplianceOSHA, UFC, IFC
  • Shipping Weight (lbs)25
  • DescriptionPolypropylene Acid Storage Cabinet, 6 X 2.5 L bottle capacity
  • Warranty1 Year
Reviews
Q&A