Scienceware Stir Bar Kit without Spinning Ring; 18 Bars
Representative image only.
Scienceware Stir Bar Kit without Spinning Ring; 18 Bars
Scienceware | Mfr: F 37184-0000| Item: 0477598
Simplify your stirring with these kits
$448.65CAD / Box of 18
BUY MORE & SAVE
  • 1-5$448.65 CAD / Box of 18
  • 6-29$403.79 CAD / Box of 18
  • 30-119$381.35 CAD / Box of 18
  • 120+$358.92 CAD / Box of 18
$448.65CAD / Box of 18
BUY MORE & SAVE
  • 1-5$448.65 CAD / Box of 18
  • 6-29$403.79 CAD / Box of 18
  • 30-119$381.35 CAD / Box of 18
  • 120+$358.92 CAD / Box of 18
Product Description
Includes
Two of 7/16" L x 3/16" Dia, Two of 1/2" L x 5/16" Dia, Two of 3/4" L x 5/16" Dia, Two of 1" L x 3/8" Dia, Two of 1-1/4" L x 3/8" Dia, Two of 1-1/2" L x 3/8" Dia, Two of 2" L x 3/8" Dia, Two of 2-1/2" L x 1/2" Dia, Two of 3" L x 1/2" Dia
Key Features
  • Chemically inert
  • Low friction
  • Leakproof PTFE coating
  • 2 year warranty
More About this Item
Kits each offer a wide variety of stir bar sizes to accomodate different glassware in the lab.
Two year warranty
Specifications
  • DescriptionWithout Spinning Ring Stir Bar Kit, 18 bars