Scienceware Serum Vial Rack for 13 to 16-mm Vials

Representative image only.

Scienceware Serum Vial Rack for 13 to 16-mm Vials

Scienceware | Mfr: 18513-1640| Item: 0673210

Withstands elevated water temperatures for incubation

$53.48CAD / Each

$53.48CAD / Each
Product Description

Key Features

  • End plate handles
  • Steam autoclavable at 121 ºC (250 ºF)
  • Holds 40 vials, 13 - 16 mm dia

More About this Item

These polypropylene racks fit most water baths for elevated temperature incubations. They feature long-lasting construction and are corrosion resistant. Height from base to shelf is 1-5/8" (41 mm). Use these racks with 10-13 mm serum vials, RIA vials, and T3-T4 reaction vials (13-16 mm).

Specifications
  • Min Tube Size (mm)13
  • Max Tube Size (mm)16
  • Tubes Held40
  • Length (in)10
  • Width (in)4 3/8
  • Height (in)3 1/2
  • Length (mm)254
  • Width (mm)111.125
  • MaterialPolypropylene(PP)
  • DescriptionSerum vial rack for 13- to 16-mm vials
Reviews
Q&A