Scienceware H36902-2006 Sterileware Bent Handle Scoop, 2 oz, 10 pack

Representative image only.

Scienceware H36902-2006 Sterileware Bent Handle Scoop, 2 oz, 10 pack

Scienceware | Mfr: H36902-2006| Item: 0626351

$57.02CAD / Pkg of 10

$57.02CAD / Pkg of 10
Product Description

Key Features

  • 3 month warranty

More About this Item

The curved handle of these scoops elevates the users hand to reduce the possibility of source contamination and exposure to hazardous substances. Made from durable polystyrene they are safe for food, drug, and cosmetic contact. A flat bottom helps keep the scoops upright allowing them to be used as a weight boat. Each individually packaged single use scoop is sterilized by gamma irradiation, lot stamped for accurate tracing, and shipped with certificate of sterilization.

3 Month Warranty
Specifications
  • Capacity (oz)2.028
  • Capacity (mL)60
  • Width (in)4 7/64
  • Width (cm)10.41
  • MaterialPolystyrene
  • SterileYes
  • DescriptionSterileware Bent Handle Scoop, 2 oz, 10 pack
  • Warranty3 month
Reviews
Q&A