Scienceware F131741008 Self Adhesive Bags, 8" x 10", 50 Pack

Representative image only.

Scienceware F131741008 Self Adhesive Bags, 8" x 10", 50 Pack

Scienceware | Mfr: F131741008| Item: 0649761

Hang anywhere for easy waste collection

$33.30CAD / Pkg of 50

$33.30CAD / Pkg of 50
Product Description

Key Features

  • 3 month warranty

More About this Item

These versatile bags can be hung from walls, cabinets or anywhere to free up valuable bench space. Simply peel the back off the self-adhesive strip and hang bag near your work area. The longer back panel makes the bag easy to open and use. Made from high-molecular weight HDPE, these white bags are steam autoclavable at 250°F (121°C). Passes both ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance tests. Maximum content weight should not exceed 3lb (1.4 kg).

3 Month Warranty
Specifications
  • MaterialHDPE
  • ColorWhite
  • Length (in)10
  • Width (in)8
  • Length (cm)25.4
  • Width (cm)20.32
  • DescriptionSelf Adhesive Bags, 8" x 10", 50 Pack
  • Warranty3 month
Reviews
Q&A