Scienceware 13201 Biohazard Autoclave Gloves, 232C max, 13" L

Representative image only.

Scienceware 13201 Biohazard Autoclave Gloves, 232C max, 13" L

Scienceware | Mfr: 13201-0000| Item: 0649640

Heat resistant to 450°F (232°C)

$51.86CAD / Pair of 1

$51.86CAD / Pair of 1

Buy It Together

Current Item

Scienceware 13201 Biohazard Autoclave Gloves, 232C max, 13" L
Scienceware 13201 Biohazard Autoclave Gloves, 232C max, 13" L
0649640

$51.86CAD / Pair of 1
+ =
Total Price 128.48 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Heat resistant to 450°F (232°C)

More About this Item

Use these soft, pliant, and all-cotton terry cloth gloves to handle hot objects fresh from the autoclave or oven. Excellent for use in any environment where heat protection is required. Bright orange for increased visibility. Machine wash, tumble dry.

Conformite Europeenne
Specifications
  • MaterialCotton
  • SizeOne size fits all
  • Length (in)13
  • Length (cm)33
  • ColorOrange
  • SterileNo
  • DescriptionBiohazard Autoclave Gloves, 232C max, 13" L
Reviews
Q&A