Sartorius Vivaspin 15 R Centrifugal Concentrators, PES Membrane 30,000 Da; 12/Pk
Representative image only.
Sartorius Vivaspin 15 R Centrifugal Concentrators, PES Membrane 30,000 Da; 12/Pk
Sartorius | Mfr: VS15RH21| Item: 3622836
Broad range of process alternatives for the filtration and concentration of biological samples
$219.54CAD / Pkg of 12
$219.54CAD / Pkg of 12
Product Description
Key Features
  • Disposable ultrafiltration devices concentrate and purify biological samples to concentrations over 100x in a single spin!
  • Concentrators feature twin vertical membranes—increased surface area provides faster filtration
  • Polyethersulfone membrane is excellent for most solutions when recovery is most important
More About this Item
This product has a modified regenerated cellulose membrane specially designed for ''sticky'' samples. This membrane has ultimate recovery at low adsorption (95 to 98%) and very small hold up volume (<20 µl). Ideal for desalting and concentration of Ig fractions.
Specifications
  • Max Molecular Weight Cut Off (Mwco) (Dalton)30000
  • Hold-Up Volume (mL)0.02
  • Max Volume Range (µL)15
  • Min Volume Range (mL)2
  • DescriptionCentrifugal Concentrators, PES Membrane 30,000 Da; 12/Pk