Saint Gobain Single-Use 3D Bioprocess Liners for Cylindrical Tanks, 200 L; 10/Pk

Representative image only.

Saint Gobain Single-Use 3D Bioprocess Liners for Cylindrical Tanks, 200 L; 10/Pk

Saint Gobain | Mfr: LR0200L| Item: 4852408

Multilayer liners provide outstanding bio and chemical compatibility, structural strength, and gas barrier—excellent for many demanding bioprocess applications

$1,885.94CAD / Pkg of 10

$1,885.94CAD / Pkg of 10
Product Description

Key Features

  • Easily integrate into your single-use process applications
  • Minimize the risk of cross-contamination and reduce cleaning time
  • Place inside tank or container—open-top bag accommodates a topdown paddle mixer
  • Bags have 5 mil (side wall) and 7 mil (bottom) film thickness
  • Free of any animal-derived components for greater process purity
  • Manufactured and packed in an ISO Class 7 certified cleanroom
  • QSR and ISO 9001:2015 compliant
  • Meet the requirements of USP Class VI and ISO 10993-3 testing
  • 200 L capacity
  • Fits tanks with 22" OD x 36"H (55.9 cm x 91.4 cm)

More About this Item

These single-use bioprocess bags feature multilayer films in different materials to provide the best biocompatibility, chemical resistance, and gas barrier to meet your bioprocessing applications. The LLDPE fluid contact layer offers outstanding biocompatibility with broad chemical compatibility for safe handling of a wide range of biopharmaceutical liquids. The EVOH middle layer ensures minimum gas permeation, ensuring the integrity of the fluid. The impact and tear resistant outside nylon layer offers outstanding durability. A multi-material tie-structure bonds the three layers, adding to the overall performance of the bag. Bags are ideal for use in buffer and media storage, bioreactor feed and harvest, and chromatography feed.

Place a liner inside a tank or containment vessel. This open-top bag accommodates a topdown paddle mixer, commonly used in upstream media preparation. Use bags in temperatures ranging from –80 to 60°C (–112 to 140°F). Bags are sterilized by gamma irradiation, ports are capped and covered, and systems are double bagged for integrity.

Dimensions given in specifications are when the liners are laid flat.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Specifications
  • MaterialLLDPE (inside), EVOH (middle), nylon (outside), and multiple materials (tie structure)
  • Capacity (Liters)200
  • ColorTranslucent
  • Length (in)42
  • Width (in)38.5
  • Height (InchShort)36.25
  • Length (cm)106.68
  • Width (cm)97.79
  • Height (cm)91.77
  • DescriptionSingle-Use 3D Bioprocess Liners for Cylindrical Tanks, 200 L; 10/Pk
Reviews
Q&A