Roscid Technologies Right Angle Bracket For Dewtran® Sensor
Representative image only.
Roscid Technologies Right Angle Bracket For Dewtran® Sensor
Roscid Technologies | Mfr: DewTran- RA-B| Item: 7893058
Accessories for DewTran® chilled mirror hygrometers
$679.97CAD / Each
$679.97CAD / Each
Product Description
Key Features
  • Sensor brackets and sample chambers make measurement easier
More About this Item
The DewTran® Series is a patented, low-cost, precision chilled mirror hygrometer. Designed for quick and accurate measurements, it reliably tracks dew point, air temperature, humidity, and barometric pressure.
Specifications
  • ColorSilver/Black
  • Compatible WithDewtran® chilled mirror hygrometers
  • DescriptionRight Angle Bracket For Dewtran® Sensor