Rex Gauge OS-2 Operation/Test Stand for Type A Durometers
Representative image only.
Rex Gauge OS-2 Operation/Test Stand for Type A Durometers
Rex Gauge | Mfr: OS-2| Item: 5979262
Convenient and accurate
$2,764.60CAD / Each
$2,764.60CAD / Each
Product Description
Key Features
  • Compliance to ASTM D-2240
  • 1 year warranty
More About this Item
Stands provide a convenient and accurate way to perform repeated hardness tests. Use stands to test pressure correctly for compliance with ASTM D-2240 and eliminate human error.
1 Year Warranty
Specifications
  • DescriptionOperation/Test Stand for Type A Durometers