ResinTech CLiR 3300 High Purity Lab Water System with 0.05 Micron Filter

Representative image only.

ResinTech CLiR 3300 High Purity Lab Water System with 0.05 Micron Filter

ResinTech | Mfr: CLS-3300| Item: 9928520

Obtain Type I, 18.2 MΩ water—on demand!

Call For Price

Call For Price

Buy It Together

Current Item

ResinTech CLiR 3300 High Purity Lab Water System with 0.05 Micron Filter
ResinTech CLiR 3300 High Purity Lab Water System with 0.05 Micron Filter
9928520

Call For Price
+ + =
Total Price 1665.01 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

0.05 micron capsule ultrafilter

Key Features

  • Meets ASTM Type I water specifications
  • Compact, flexible configurations save counter space
  • Built-in timer prevents excess wear
  • 2.5 LPM flowrate
  • Built-in leak detection
  • Intuitive user interface
  • Easy cartridge replacement
  • Low operating costs
  • Large capacity, low-cost consumable cartridges
  • Genuine ResinTech media
  • Made in the USA
  • 0.05 micron capsule ultrafilter removes bacteria
  • 1 year warranty

More About this Item

CLiR high-purity water systems features continual recirculation to ensures constant ultra-pure water for use in your lab. Choose from systems offering up to 4.0 liters of 18.2 megohm water per minute. Standard features include a built-in pressure regulator, high-capacity cartridges for deionizing the water, and a POU capsule capsule filter for removal of particulates and unwanted bacteria from the water as it is being dispensed through the outlet valve.

Multiple sensors continuously monitor the system and final water quality to ensure trouble-free operation. A digital resistivity monitor with alarm LED set points provides system feedback performance. The large volume cartridges require less change-outs, offering you lower operating costs. [Note: The feed water quality will determine the necessary pre-treatment. For optimum performance, the water system may require reverse osmosis (99284-51), distillation, or deionizing pretreatment.]

Small footprints and flexible mounting options (counter, wall, and under-sink) allow you to maximize valuble counter space. Consumables are simple to replace and require no tools (Cartridges sold separately.). Dispensing gun and secondary auxiliary side port are available upon request. Additional options are available for special applications to include 185/254 nm UV, and or ultrafiltration (UF).
Some CLiR system have been pre-configured into four application specific models with benchtop or remote dispesing options:
For Basic chemistry, academia, IC, and buffers: CLiR 5100 (models 99285-31 and -32)
For HPLC, GC-MS, and trace organics: CLiR 5200 (models 99285-35 and -36 with UV)
For Life science, cell culture, and microbiology: CLiR 5300 (models 99285-33 and -34 with UF)
For DNA Sequencing, PCR, and electrophoresis: CLiR 5400 (models 99285-37 and -38 with UV and UF)

A complete package requires a system, conditioning cartridge, polishing cartridge, and final filter.
1. Choose CLiR 5000 base system
2. Choose conditioning cartridge 99285-45 for RO water feed, or 99285-46 for SDI/tap water feed
3. Choose polishing cartridge 99285-43 for standard results, or 99285-44 for additional organics removal
4. Choose final filter 99285-27 for up to 0.2µm pariculate removal, or 99285-47 to also include endotoxin removal

Options (must be ordered with unit, factory installation required):
Item 99285-39 - adds in flow totalizer function, viewable on display
Item 99285-40 - adds in a direct feed port
Item 99285-41 - adds in a recirculating dispensing gun (for units without a remote dispense function)
Item 99285-42 - adds in a wall mounting bracket to clear up bench space"

1 Year Warranty
Specifications
  • ASTM Water GradeType I
  • Max Flow Rate (L/min)4
  • Number of Holders3
  • Inlet Port3/8" OD tubing
  • With UV LampNo
  • With UltrafilterYes
  • Type I Water (Resistivity)18.2 MΩ
  • Type I Water (TOC)5 to 10 ppb
  • Type I Water (Bacteria)< 1 CFU/mL
  • Type I Water (Endotoxins)< 0.03 EU/mL
  • Max Inlet Water Temperature (° F)100
  • Max Inlet Water Temperature (° C)30
  • Min Inlet Pressure (PSI)20
  • Max Inlet Pressure (PSI)60
  • Min Inlet Pressure (bar)1.4
  • Max Inlet Pressure (bar)4.1
  • Power (VAC)120
  • Height (in)25
  • Depth (in)8.5
  • Width (in)23
  • Height (cm)64
  • Depth (cm)22
  • Width (cm)59
  • DescriptionHigh Purity Water System with 0.05 Micron Capsule Ultrafilter
  • Warranty1 year
Reviews
Q&A