Rapid Surface Hygiene Kit, 50 tests/kit
Representative image only.
Rapid Surface Hygiene Kit, 50 tests/kit
| Item: 1404030
Detects invisible food residue without an instrument -in only 10 to 60 seconds
$289.15CAD / Each
$289.15CAD / Each
Buy It Together
Current Item
Rapid Surface Hygiene Kit, 50 tests/kit
Rapid Surface Hygiene Kit, 50 tests/kit
1404030
$289.15CAD / Each
+
GVS <i>Escherichia coli</i> SwabCheck™ Swabs, 4 mL; 125/PK
7890945
$1,330.47CAD / Pkg of 125
=
Total Price 1619.62 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
VERIcleen is sensitive to food residues including meat, dairy, bakery, beverages, eggs, fish, etc. It is an inexpensive rapid hygiene program to the food service, retail and institutional industries. Applications include HACCP programs, hand washing hygiene compliance, and overall sanitation monitoring.
Application is extremely simple, just spray to wet dry surface and swab. A ''clean surface'' shows no purple until after 3 minutes. A ''Not clean'' surface shows purple in 10 to 60 seconds.
Each kit includes 50 tests in a light-tight snap-top container, 1 bottle of wetting liquid with atomizer, instruction leaflet, and desiccant packet (if desiccant inside the packet has turned from blue to pink, the strips have been stored improperly, and may be invalid).
Specifications
  • Test TypeFood Residues (meat, dairy, bakery, beverages, eggs, fish, etc.)
  • Number of Tests Included50
  • DescriptionRapid Surface Hygiene Kit, 50 tests/kit