Radiation Alert Personal Radiation Dosimeter, 0.1 to 2.0 Msv
Representative image only.
Radiation Alert Personal Radiation Dosimeter, 0.1 to 2.0 Msv
Radiation Alert | Mfr: PEN2MSV| Item: 8192215
Direct-reading dosimeter measures gamma and x-ray radiation exposure
$270.74CAD / Each
BUY MORE & SAVE
  • 1-4$270.74 CAD / Each
  • 5+$243.67 CAD / Each
$270.74CAD / Each
BUY MORE & SAVE
  • 1-4$270.74 CAD / Each
  • 5+$243.67 CAD / Each
Buy It Together
Current Item
Radiation Alert Personal Radiation Dosimeter, 0.1 to 2.0 Msv
Radiation Alert Personal Radiation Dosimeter, 0.1 to 2.0 Msv
8192215
$270.74CAD / Each
+ =
Total Price 787.90 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Rugged charger requires no batteries — simply squeeze the trigger of this piezoelectric generator to move the hairline fiber inside the dosimeter back to zero
  • Clamping mechanism automatically locks the dosimeter in place
  • Charger is easy to operate and requires only few squeezes of the trigger for each dosimeter
  • 2 year warranty
More About this Item
Personal, pen-style dosimeters clip easily to your pocket or a specific object to measure personal accumulated dose or quantity of gamma and x-ray radiation. Dosimeter is a conductive-fiber electroscope with a thin wall to allow penetration of radiation. Inside the dosimeter, the fiber moves across a predetermined scale to display the level of radiation exposure. Readings are easily displayed by holding the dosimeter up to a light source to view the scale — immediate results without external lab analysis. Order handheld charger (sold separately) to zero dosimeters and reset scale for repeated use.
Two year warranty
Specifications
  • Range0.1 to 2 mSv
  • Accuracy±10% of true dose for Cs-137 or Co-60 gamma
  • Resolution0.1 mSv
  • DescriptionPersonal Radiation Dosimeter, 0.1 to 2.0 Msv