Qsonica 4422 Ultrasonic Cell Disruptor Microtip Probe, Stepped 1/8" (3.2 mm) dia, 0.5-15 mL
Representative image only.
Qsonica 4422 Ultrasonic Cell Disruptor Microtip Probe, Stepped 1/8" (3.2 mm) dia, 0.5-15 mL
Qsonica | Mfr: 4422| Item: 0471265
Microtip Probes for use with Cole-Parmer and Qsonica Q500 and Q700 Sonicators
$500.95CAD / Each
$500.95CAD / Each
Buy It Together
Current Item
Qsonica 4422 Ultrasonic Cell Disruptor Microtip Probe, Stepped 1/8" (3.2 mm) dia, 0.5-15 mL
Qsonica 4422 Ultrasonic Cell Disruptor Microtip Probe, Stepped 1/8" (3.2 mm) dia, 0.5-15 mL
0471265
$500.95CAD / Each
+ + =
Total Price 12274.07 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Microtips screw into the threaded end of standard ½" probe
  • 1 year warranty
More About this Item
Probes effectively process small samples and attach to the standard 1/2"-dia probe horn. The stepped microtip probe (04711-93) attaches directly to the converter and is ideal for reaching into long, thin tubes.