Qsonica 4408 Replacement Tips for Standard Probe, 1" (25.4 mm)
Representative image only.
Qsonica 4408 Replacement Tips for Standard Probe, 1" (25.4 mm)
Qsonica | Mfr: 4408| Item: 0471274
Microtip Probes for use with Cole-Parmer and Qsonica Q500 and Q700 Sonicators
$110.50CAD / Each
$110.50CAD / Each
Buy It Together
Current Item
Qsonica 4408 Replacement Tips for Standard Probe, 1" (25.4 mm)
Qsonica 4408 Replacement Tips for Standard Probe, 1" (25.4 mm)
0471274
$110.50CAD / Each
+ + =
Total Price 11883.62 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Microtips screw into the threaded end of standard ½" probe
  • 1 year warranty
More About this Item
Probes effectively process small samples and attach to the standard 1/2"-dia probe horn. The stepped microtip probe (04711-93) attaches directly to the converter and is ideal for reaching into long, thin tubes.