Qsonica 4220 Sonicator Probe with Replaceable Tip; 1/2" Dia
Representative image only.
Qsonica 4220 Sonicator Probe with Replaceable Tip; 1/2" Dia
Qsonica | Mfr: 4220| Item: 0471266
$1,375.41CAD / Each
$1,375.41CAD / Each
Buy It Together
Current Item
Qsonica 4220 Sonicator Probe with Replaceable Tip; 1/2" Dia
Qsonica 4220 Sonicator Probe with Replaceable Tip; 1/2" Dia
0471266
$1,375.41CAD / Each
+ =
Total Price 12518.66 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Replacement Probe selection is determined by sample volume, sample type, and the desired intensity. As tip diameter increases, the intensity/amplitude decreases. Tips erode over time and replacement tips are available for order.
1 Year Warranty
Specifications
  • DescriptionSonicator Probe with Replaceable Tip; 1/2" Dia