QRP VHC12 SMALL Vinyl Cleanroom Gloves, small, 100/bag
Representative image only.
QRP VHC12 SMALL Vinyl Cleanroom Gloves, small, 100/bag
QRP | Mfr: VHC12 SMALL| Item: 8658020
Perfect for pharmaceuticals, electronics, and cleanroom food processing
$66.16CAD / Bag of 100
BUY MORE & SAVE
  • 1-9$66.16 CAD / Bag of 100
  • 10+$59.54 CAD / Bag of 100
$66.16CAD / Bag of 100
BUY MORE & SAVE
  • 1-9$66.16 CAD / Bag of 100
  • 10+$59.54 CAD / Bag of 100
Product Description
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Gloves feature a micro-textured surface for enhanced gripping. Latex-free, 100% PVC co-polymer is naturally antistatic with a resistivity of 1010 ohms. Beaded cuff for additional strength. Ambidextrous. Packed 100 gloves per polybag.
Compliance: ISO Class 5, USDA, and IEST-RP-CC-005.2.
Specifications
  • Thickness (mil)6
  • MaterialVinyl
  • SizeSmall
  • Length (in)12
  • Length (cm)30.5
  • ColorWhite
  • CuffBeaded
  • ComplianceISO Class 5; USDA; IEST-RP-CC-005
  • SterileNo
  • DescriptionVinyl Cleanroom Gloves, small, 100/bag