QRP 7C LARGE Latex Antistatic Finger Cots, large
Representative image only.
QRP 7C LARGE Latex Antistatic Finger Cots, large
QRP | Mfr: 7C LARGE| Item: 8628302
Minimize charge generation
$66.16CAD / Pkg of 720
BUY MORE & SAVE
  • 1-9$66.16 CAD / Pkg of 720
  • 10+$59.54 CAD / Pkg of 720
$66.16CAD / Pkg of 720
BUY MORE & SAVE
  • 1-9$66.16 CAD / Pkg of 720
  • 10+$59.54 CAD / Pkg of 720
Product Description
Key Features
  • Anti-Static Vacuum Sealed Latex Finger Cots ISO 5 (Class 100)
  • 3 month warranty
More About this Item
Latex finger cots are washed, powder-free, and treated with an anti-static agent. Use to handle Class II static-sensitive devices with thresholds above 1000 V. Average surface resistivity is 5 X 1012 ohms. Each pack is double-bagged in ESD-safe packaging to prevent charge generation.
Compliance:
ISO Class 5.
ASTM and IEST-RP-CC 005.3 standards.
ESD Safe to ANSI/ESD S20.20-2014
3 Month Warranty
Specifications
  • Thickness (mil)3
  • MaterialLatex
  • SizeLarge
  • ColorPink
  • ComplianceISO Class 5
  • SterileNo
  • DescriptionLatex Antistatic Finger Cots, large