QLA Porous Filters, 4 µm, UHMW PE, 1/8" ID, Hanson compatible; 100/PK

Representative image only.

QLA Porous Filters, 4 µm, UHMW PE, 1/8" ID, Hanson compatible; 100/PK

| Mfr: FIL004-HR-A| Item: 4130891

Achieve reproducibility and consistency from test to test with QLA dissolution filters

$280.68CAD / Pkg of 100

$280.68CAD / Pkg of 100
Product Description

Key Features

  • Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal Made of Ultra High Molecular Weight (UHMW) polyethylene
  • 4 µm pore size
  • 100 per pack

More About this Item

QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filters are completely inert to most compounds and provide a good flow rate.

.

Specifications
  • MaterialUHMW Polyethylene
  • Pore Size (micron)4
  • Internal Diameter (mm)3.2
  • DescriptionPorous Filters, 4 µm, UHMW PE, 1/8" ID, Hanson compatible; 100/PK
Reviews
Q&A