Pyrex 9985-150 Brand 9985 watch glass; 150 mm, case of 72
Representative image only.
Pyrex 9985-150 Brand 9985 watch glass; 150 mm, case of 72
Pyrex | Mfr: 9985-150| Item: 3450726
$1,013.69CAD / Case of 72
BUY MORE & SAVE
  • 1-4$1,013.69 CAD / Case of 72
  • 5-9$912.32 CAD / Case of 72
  • 10+$861.64 CAD / Case of 72
$1,013.69CAD / Case of 72
BUY MORE & SAVE
  • 1-4$1,013.69 CAD / Case of 72
  • 5-9$912.32 CAD / Case of 72
  • 10+$861.64 CAD / Case of 72
Product Description
More About this Item
These watch glasses provide high chemical durability, preventing contamination of solutions. They are modeled to the same radius of curvature for convenient stacking. The heavy wall an uniform fire-polished edge provide mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected to occur.
Specifications
  • MaterialGlass
  • Fits Bottle / Beaker Size (mL)2000
  • AutoclavableYes
  • Outside Diameter (OD) (cm)15
  • DescriptionBrand 9985 watch glass; 150 mm, case of 72