Pyrex 4320-500 Brand 4320 round-bottom flask; 500 mL, pack of 2
Representative image only.
Pyrex 4320-500 Brand 4320 round-bottom flask; 500 mL, pack of 2
Pyrex | Mfr: 4320-500| Item: 3455304
Boiling, round bottom, short neck, joint
$187.12CAD / Pkg of 2
BUY MORE & SAVE
  • 1-4$187.12 CAD / Pkg of 2
  • 5-9$168.41 CAD / Pkg of 2
  • 10+$159.05 CAD / Pkg of 2
$187.12CAD / Pkg of 2
BUY MORE & SAVE
  • 1-4$187.12 CAD / Pkg of 2
  • 5-9$168.41 CAD / Pkg of 2
  • 10+$159.05 CAD / Pkg of 2
Buy It Together
Current Item
Pyrex 4320-500 Brand 4320 round-bottom flask; 500 mL, pack of 2
Pyrex 4320-500 Brand 4320 round-bottom flask; 500 mL, pack of 2
3455304
$187.12CAD / Pkg of 2
+ + =
Total Price 267.57 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These boiling flasks with short neck are carefully designed and manufactured from borosilicate glass. Uniform wall thickness, a characteristic of all round-bottom, Pyrex glassware, provides a balance between expansion and mechanical strength for greater shock resistance and versatility. Use boiling flasks for a variety of mixing, heating, and boiling applications commonly encountered in most laboratory work. The normal working temperature is 230°>C. Includes a Ts joint.
Specifications
  • MaterialGlass
  • Capacity (mL)500
  • Overall Height (cm)16
  • Joint Size Number24/40
  • Base Diameter (cm)10.2
  • DescriptionBrand 4320 round-bottom flask; 500 mL, pack of 2