Pomona Electronics 72929-20-2 Mini Smd Grabber Test Clip with Lead, 4mm, Jack Red

Representative image only.

Pomona Electronics 72929-20-2 Mini Smd Grabber Test Clip with Lead, 4mm, Jack Red

Pomona Electronics | Mfr: 72929-20-2| Item: 2000971

Available for sale only in the United States and Canada

$47.03CAD / Each

$47.03CAD / Each
Product Description

Key Features

  • Designed for easy attachment to SMD chips with pitches down to 0.032"
  • 3 month warranty

More About this Item

Test Clips

Mini SMD Grabber Test Clips
Insulated shaft can be bent up to 36 degrees for versatile attachment in awkward locations or close proximity to other clips. Rotating jaws increase versatility in connection and stacking. Thin (3mm) profile allows close placement to adjacent clips.
Available for sale in the United States and Canada only.

3 Month Warranty
Specifications
  • DescriptionMini Smd Grabber Test Clip with Lead, 4mm, Jack Red
  • Warranty3 month
Reviews
Q&A