Pomona Electronics 72928 SMD Multi-Use Test Kit, Model
Representative image only.
Pomona Electronics 72928 SMD Multi-Use Test Kit, Model
Pomona Electronics | Mfr: 72928| Item: 2303172
Available for sale only in the United States and Canada.
$359.65CAD / Each
$359.65CAD / Each
Product Description
Includes
Kit contains: (2) Micro SMD Grabber Test Clips (black, red); (2) Mini-alligator clip with 2mm jack (black, red); (2) SMD test probe, spring-set tip (black, red); (2) Mini SMD Grabber Test Clips (black, red); (2) 4mm plug / 2mm plug adapter (black, red); B
Key Features
  • SMD micro and mini grabbers attach to chip leads with pitches down to 0.008 (0.20 mm)
  • Includes all adapters and leads necessary for attachment to standard bench or handheld DMM
  • Probe tips are sharp and spring-loaded for solid and steady contact with SMD test points
  • Compatible with all multimeters that accept standard sheathed banana plugs
  • Test accessories conveniently store in four-fold nylon pouch
Specifications
  • DescriptionSMD Multi-Use Test Kit, Model