Pomona Electronics 3781 Test Clip Patch Cord, 24", Red

Representative image only.

Pomona Electronics 3781 Test Clip Patch Cord, 24", Red

Pomona Electronics | Mfr: 3781-24-2| Item: 2303188

Ideal for providing temporary jumper wires

$13.19CAD / Each

$13.19CAD / Each
Product Description

Key Features

  • Available in red, 24" length

More About this Item

Provides excellent connector to terminals and wire wrap posts.

Available for sale only in the United States and Canada.

Specifications
  • DescriptionPatch Cord, 24 in Red
Reviews
Q&A