Pomona Electronics 3781 Test Clip Patch Cord, 24", Black

Representative image only.

Pomona Electronics 3781 Test Clip Patch Cord, 24", Black

Pomona Electronics | Mfr: 3781-24-0| Item: 2303187

Ideal for providing temporary jumper wires

$13.19CAD / Each

$13.19CAD / Each
Product Description

Key Features

  • Available in black, 24" length

More About this Item

Provides excellent connector to terminals and wire wrap posts.

Available for sale only in the United States and Canada.

Specifications
  • DescriptionOptional Patch Cord, 24in Black
Reviews
Q&A