PolyScience External Circulation Tubing, Buna-N, 13-mm ID; 1-meter
Representative image only.
PolyScience External Circulation Tubing, Buna-N, 13-mm ID; 1-meter
PolyScience | Mfr: 060308| Item: 0126740
$48.62CAD / Yard of 1
$48.62CAD / Yard of 1
Product Description
Key Features
  • 2-1/2 Years Parts and Labor Warranty
More About this Item
For use in applications requiring external circulation, Buna-N is a synthetic rubber capable of handling temperatures from -40 to 120°C. Order multiple 1-meter segments for a continuous length.
Compatible (without additional fittings) with the following PolyScience models.
LM series benchtop chiller
LS series benchtop chiller
MM series benchtop chiller
6000 series chiller
3370 liquid-to-air cooler
4100 liquid-to-liquid cooler
2 and a Half Year Warranty
Specifications
  • Description060308 External Circulation Tubing, Buna-N, 13-mm ID; 1-meter L 01267-40
  • Warranty2-1/2 Years Parts and Labor