PIP 98-740-S Nylon Inspection Gloves, 40 Denier, Small, 12 pairs/pk
Representative image only.
PIP 98-740-S Nylon Inspection Gloves, 40 Denier, Small, 12 pairs/pk
PIP | Mfr: 98-740-S| Item: 8636909
Low—lint nylon is an excellent choice for inspection of electronic components
$23.13CAD / Dozen of 12
BUY MORE & SAVE
  • 1-9$23.13 CAD / Dozen of 12
  • 10+$20.82 CAD / Dozen of 12
$23.13CAD / Dozen of 12
BUY MORE & SAVE
  • 1-9$23.13 CAD / Dozen of 12
  • 10+$20.82 CAD / Dozen of 12
Product Description
More About this Item
Low-lint nylon inspection gloves are ideal for areas that are sensitive to lint particles often created by cotton gloves. These affordable inspection gloves are washable and reusable. Gloves are lightweight for extra sensitivity. Ambidextrous.
Economy gloves are constructed using 40 denier tricot nylon in a two-piece pattern and feature rolled cuffs.
Specifications
  • MaterialNylon
  • SizeSmall
  • ColorWhite
  • CuffRolled
  • SterileNo
  • DescriptionNylon Inspection Gloves, 40 Denier, Small, 12 pairs/pk