PIP 98-740-L Nylon Inspection Gloves, 40 Denier, Large, 12 pairs/pk
Representative image only.
PIP 98-740-L Nylon Inspection Gloves, 40 Denier, Large, 12 pairs/pk
PIP | Mfr: 98-740-L| Item: 8636913
Low—lint nylon is an excellent choice for inspection of electronic components
$26.37CAD / Dozen of 12
BUY MORE & SAVE
  • 1-9$26.37 CAD / Dozen of 12
  • 10+$23.73 CAD / Dozen of 12
$26.37CAD / Dozen of 12
BUY MORE & SAVE
  • 1-9$26.37 CAD / Dozen of 12
  • 10+$23.73 CAD / Dozen of 12
Product Description
More About this Item
Low-lint nylon inspection gloves are ideal for areas that are sensitive to lint particles often created by cotton gloves. These affordable inspection gloves are washable and reusable. Gloves are lightweight for extra sensitivity. Ambidextrous.
Nylon Inspection Gloves are lightweight for extra sensitivity.
Low-lint gloves are ideal for areas that are sensitive to lint particles often created by cotton gloves. Two-piece construction makes these unhemmed gloves comfortable enough for all day wear.
Specifications
  • MaterialNylon
  • SizeLarge
  • ColorWhite
  • CuffRolled
  • SterileNo
  • DescriptionNylon Inspection Gloves, 40 Denier, Large, 12 pairs/pk