Orochem Reliasil™ HPLC Column, Silica, 5 µm, 100 mm L x 3.0 mm ID, 90Å

Representative image only.

Orochem Reliasil™ HPLC Column, Silica, 5 µm, 100 mm L x 3.0 mm ID, 90Å

Orochem | Mfr: R5DG-120| Item: 0648671

Rugged design with enhanced chemical stability and efficiency

$1,012.22CAD / Each

$1,012.22CAD / Each
Product Description

Key Features

  • Excellent mechanical and chemical stability
  • Ideal for reverse phase separations of peptides, pharmaceuticals, and derivatized amino acids
  • Suitable for Silica phase
  • Silica column dimensions are 100 mm long by 5 mm diameter

More About this Item

The Orochem Reliasil HPLC columns are designed for high efficiency (high plate count) and general utility. The proprietary silica gel manufacturing process yields a Gaussian particle size distribution, narrow pore-size distribution, and strong resistance to high pH. All columns feature a 190 m²/g surface area and 0.55 pore volume.Standard columns feature a pore size of 90 Å.

Specifications
  • PhaseSilica
  • Column Length (mm)100
  • Column Inside Diameter (ID) (mm)3
  • Column Particle Size (µm)5
  • DescriptionReliasil HPLC Column, Silica, 100 x 3 mm, 5 µm
Reviews
Q&A