Orochem Reliasil™ HPLC Column, Silica, 3 µm, 150 mm L x 4.6 mm ID, 90Å

Representative image only.

Orochem Reliasil™ HPLC Column, Silica, 3 µm, 150 mm L x 4.6 mm ID, 90Å

Orochem | Mfr: R3FI-100| Item: 0648672

Rugged design with enhanced chemical stability and efficiency

$1,046.11CAD / Each

$1,046.11CAD / Each
Product Description

Key Features

  • Excellent mechanical and chemical stability
  • Ideal for reverse phase separations of peptides, pharmaceuticals, and derivatized amino acids
  • Suitable for Silica phase
  • Silica column dimensions are 150 mm long by 3 mm diameter

More About this Item

The Orochem Reliasil HPLC columns are designed for high efficiency (high plate count) and general utility. The proprietary silica gel manufacturing process yields a Gaussian particle size distribution, narrow pore-size distribution, and strong resistance to high pH. All columns feature a 190 m²/g surface area and 0.55 pore volume.Standard columns feature a pore size of 90 Å.

Specifications
  • PhaseSilica
  • Column Length (mm)150
  • Column Inside Diameter (ID) (mm)4.6
  • Column Particle Size (µm)3
  • DescriptionReliasil HPLC Column, Silica, 150 x 4.6 mm, 3 µm
Reviews
Q&A