Ohaus Genie SI-1100 Roto-Shake Rotator/Rocker; 120 VAC

Representative image only.

Ohaus Genie SI-1100 Roto-Shake Rotator/Rocker; 120 VAC

Ohaus | Mfr: SI-1100| Item: 5160300

Multi-action platform performs rocking or rotating motions. Magnetic stainless steel strips and spring-clip mounting plates offer a variety of configurations for different applications. Wide ambient range is ideal for use in cold rooms and incubators.

$2,239.55CAD / Each

$2,239.55CAD / Each

Buy It Together

Current Item

Ohaus Genie SI-1100 Roto-Shake Rotator/Rocker; 120 VAC
Ohaus Genie SI-1100 Roto-Shake Rotator/Rocker; 120 VAC
5160300

$2,239.55CAD / Each
+
Ohaus Genie SI-1170 Roto-Shake Stackable Tray
5160386

$257.47CAD / Each
=
Total Price 2497.02 ADD 2 ITEMS TO CART
Product Description

Includes

one plate each for 12 x 10 to 13 mm, 6 x 15 to 17 mm, and 3 x 28 to 30 mm dia tubes; four magnetic stainless steel mounting strips for 6" x 6" plastic bags; one non-skid metal rocker platform (12" x 12").

Key Features

  • 2 year warranty

More About this Item

Now you can provide a host of shaking options for your applications with the addition of this all-in-one shaker unit. It easily changes from rocking to rotating, and the magnetic platform may be removed and replaced with a 3-D orbital shaking platform.
A simple mechanical adjustment changes the rocker/rotator function. The moving component of this shaker is a two-sided 8"W x 13.5"L stainless steel platform. This accommodates a wide range of attachments that mount in a variety of configurations for different applications.
Use magnetic SS strips to mount membrane-based blot assays/washings directly on platform. The non-skid metal rocker platform attachment for trays, and the autoclavable spring-clip mounting plates for three different size tubes fasten magnetically to the platform.
The stainless steel platform is easy to clean and decontaminate. Because shaker maintains set speeds between 0 and 38°C, it is suitable for use in cold rooms and incubators. For greater capacity, order expansion kits separately.

As of September 2025 Scientific Industries sold the Vortex Genie product line to Ohaus

UL RecognizedETL ListedTwo year warranty
Specifications
  • Shaking Angle±10° from horizontal
  • Min Speed (rpm)5
  • Max Speed (rpm)35
  • Tube Size12 tubes of 10 to 13 mm diameter; 6 tubes of 15 to 17 mm diameter; 3 tubes of 28 to 30 mm diameter; 2 plastic bags of 6" x 6"; 1 tray of 12" x 12"
  • Platform Length (in)13.5
  • Platform Width (in)8
  • Platform Length (cm)34.3
  • Platform Width (cm)20.3
  • Width (in)10.5
  • Height (in)20
  • Depth (in)9.9062
  • Width (cm)26.67
  • Height (cm)50.8
  • Depth (cm)25.161748
  • Max Operating Temperature (° F)100
  • Max Operating Temperature (° C)38
  • Power (VAC)120
  • Power (Hz)60 Hz
  • Power (amps)5
  • DescriptionRoto-Shake Rotator/Rocker; 120 VAC
  • Warranty2 year
Reviews
Q&A