Ohaus E51LDMP03 Endeavor Standard Duty Orbital Digital Microplate Shaker with 3mm Orbit, 100 to 1200 rpm

Representative image only.

Ohaus E51LDMP03 Endeavor Standard Duty Orbital Digital Microplate Shaker with 3mm Orbit, 100 to 1200 rpm

Ohaus | Mfr: E51LDMP03| Item: 5302683

Triple eccentric drive provides precise, reliable sample mixing

$1,471.62CAD / Each

$1,471.62CAD / Each

Buy It Together

Current Item

Ohaus E51LDMP03 Endeavor Standard Duty Orbital Digital Microplate Shaker with 3mm Orbit, 100 to 1200 rpm
Ohaus E51LDMP03 Endeavor Standard Duty Orbital Digital Microplate Shaker with 3mm Orbit, 100 to 1200 rpm
5302683

$1,471.62CAD / Each
+
Ohaus Micro Tube Rack for use with Shaker Platforms
5302711

$306.91CAD / Each
=
Total Price 1778.53 ADD 2 ITEMS TO CART
Product Description

Includes

Aluminum tray with hardware to secure microplates and deepwell plates.

Key Features

  • Remote function permits PC control and data transfer
  • Bright, backlit LCD display clearly shows speed and time
  • Durable chemical-resistant polymer housing
  • Easy-to-clean aluminum standard tray
  • IP21 ingress protection
  • Speeds up to 1200 rpm
  • 2 year warranty

More About this Item

These light duty orbital digital shakers provide versatility for different shaking requirements. They are compatible with use in both incubators and cold rooms, ensuring flexibility across a wide range of environmental conditions. Applications include ELISA assays, cell cultures, protein studies, bacterial and yeast cultures, and DNA studies.

The compact design of the shakers saves you valuable bench space, while the advanced microprocessor control optimizes speed ramp-up to avoid splashing. The triple eccentric drive and maintenance-free brushless DC motor offers stable, accurate, and long-term shaking. A simple on-board menu allows you to set, store, and recall multi-step programs, while the RS232 interface provides two-way communication for data logging and remote control of the unit.

Safety features include an integrated overload and unbalanced detection to protect samples from potential damage, ensuring the reliability of experimental results. A visual and audio alarm system alerts you when the timer counts down to zero or an error occurs.

The easy-to-clean aluminum tray features 11 mounting points for various accessories allowing you to customize your shaker with tube racks, flask clamps, and a non-slip rubber mat.

The shakers are available in three models: 3 mm orbit for gentle mixing, 20 mm orbit for more vigorous applications, and the dedicated microplate model.

Two year warrantyConformite EuropeenneCanadian Standards Association
Specifications
  • Orbit Size (in)0.1
  • Orbit Size (mm)3
  • Min Speed (rpm)100
  • Max Speed (rpm)1200
  • Speed Accuracy (rpm)Above 100 rpm ± 2% of set speed, Below 100 rpm ±2% rpm
  • Max Load (lbs)8.8
  • Max Load (kg)4
  • Platform Flask Capacity4 microplates
  • Platform Length (in)11
  • Platform Width (in)7.75
  • Platform Length (cm)28
  • Platform Width (cm)19.7
  • Width (in)10.3
  • Height (in)4.7
  • Depth (in)16.5
  • Width (cm)26.2
  • Height (cm)11.9
  • Depth (cm)42
  • Min Operating Temperature (° F)14
  • Max Operating Temperature (° F)104
  • Min Operating Temperature (° C)-10
  • Max Operating Temperature (° C)40
  • TimerYes
  • Timer RangeUp to 99 hours 59 minutes
  • DisplayLCD
  • Housing MaterialChemical-resistant polymer
  • Power (VAC)24
  • Power (amps)1.25
  • DescriptionEndeavor Standard Duty Orbital Digital Microplate Shaker with 3mm Orbit, 100 to 1200 rpm
  • Warranty2 year
Reviews
Q&A