Environmental Express Oakton CTSTestr™ 50C Cup-Style Waterproof Pocket Conductivity/TDS/Salinity Tester, Premium 50 Series

Representative image only.

Environmental Express Oakton CTSTestr™ 50C Cup-Style Waterproof Pocket Conductivity/TDS/Salinity Tester, Premium 50 Series

Premium multi parameter pocket testers for field and lab testing

$362.45CAD / Each

for an additional
$153.97CAD / Each
for an additional
$276.26CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$362.45CAD / Each

Buy It Together

Current Item

Environmental Express Oakton CTSTestr™ 50C Cup-Style Waterproof Pocket Conductivity/TDS/Salinity Tester, Premium 50 Series
Environmental Express Oakton CTSTestr™ 50C Cup-Style Waterproof Pocket Conductivity/TDS/Salinity Tester, Premium 50 Series
3563465

$362.45CAD / Each
+ + + + + +
Environmental Express Buffer Solution, pH 10.01; 500 mL
0065408

$29.62CAD / Pint of 1
+ + + =
Total Price 1500.06 ADD 10 ITEMS TO CART
Product Description

Includes

4 AAA batteries.

Key Features

  • Available in pH, Conductivity, TDS, Salinity, and ORP models
  • Replaceable, double-junction sensors for durability and longevity against aggressive samples
  • Four-button keypad for technician level use
  • Leakproof cap functions as a sample cup, hydrating cup, and stabilizing base to stand the meter upright
  • 1 year warranty

More About this Item

Our most advanced line of pocket testers, the Oakton 50 Series provides single and multimeter options for measuring pH, Conductivity, TDS, Salinity, and ORP. Replaceable double junction sensors provide durability and longevity against highly caustic, volatile or hazardous samples in both lab and field applications. Waterproof and designed to float in water, they are ideal for field-based testing environments – with a convenient loop for security line attachment.

50 series pocket meters include a replaceable double-junction electrode sensor that is designed for more aggressive applications, including wastewater and solutions containing sulfides, heavy metals, and TRIS. The electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage and allow for easy maintenance and replacement. A Valox sensor housing resists most chemicals.

An easy-to-use four-button keypad allows for technician level use and measurement with quick access to a variety of functions. Up to 3 calibration points with auto-buffer recognition at both USA and NIST standard buffers. Additional features include an automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.


1 Year WarrantyConformite EuropeenneIP 67ISO 9001:2008 Certified Supplier
Specifications
  • Meter TypeTester
  • Display typeLCD
  • CalibrationUp to 3 points
  • Min Conductivity0.1 uS
  • Max Conductivity20.00 mS
  • Conductivity Resolution0.1 uS, 1 uS, 0.01 mS
  • Conductivity Accuracy±1% full scale
  • Min TDS0.1 ppm
  • Max TDS10.00 ppt
  • TDS Resolution0.1 ppm, 1 ppm, 0.01 ppt
  • TDS Accuracy±1% full scale
  • Min Salinity0.1 ppt
  • Max Salinity10.00 ppt
  • Salinity Resolution0.1 ppt
  • Salinity Accuracy±1% full scale
  • Min Temperature (° C)0
  • Max Temperature (° C)60
  • Min Temperature (° F)32
  • Max Temperature (° F)140
  • Temperature Resolution0.1°C / 0.1°F
  • Temperature Accuracy±0.5°C, ±0.9°F
  • Temperature compensationYes, ATC
  • WaterproofYes
  • Battery4 AAA (included)
  • Battery life (hr)250
  • DescriptionCup-Style Waterproof Pocket Conductivity/TDS/Salinity Tester, Premium 50 Series
  • Warranty1 year
Reviews
Q&A