Novasina ClimMate nSens-HT-ENS Replacement Humidity and Temperature Probe for ClimMate Portable Water Activity Meter
Representative image only.
Novasina ClimMate nSens-HT-ENS Replacement Humidity and Temperature Probe for ClimMate Portable Water Activity Meter
Novasina | Mfr: 260 1171| Item: 3791062
Keep your Novasina ClimMate Portable Water Activity Meter producing optimal results
$1,433.61CAD / Each
$1,433.61CAD / Each
Buy It Together
Current Item
Novasina ClimMate nSens-HT-ENS Replacement Humidity and Temperature Probe for ClimMate Portable Water Activity Meter
Novasina ClimMate nSens-HT-ENS Replacement Humidity and Temperature Probe for ClimMate Portable Water Activity Meter
3791062
$1,433.61CAD / Each
+ =
Total Price 1972.13 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
nCap-PS protective filter cap.
Key Features
  • Choose from a wide range of accessories including humidity standards, probes, and more
More About this Item
These replacement accessories are compatible with Novasina ClimMate Portable Water Activity meter, model 37910-38. Meter sold separately.
This replacement digital humidity and temperature sensor features electrolytic humidity sensor and NTC temperature measurement technology. The adjustment points are stored directly in the sensor. This probe is compatible with the ClimMate water activity meter, model 37910-38, meter sold separately.
Cole-Parmer cannot sell this product in China, Japan, Germany, France, Italy, Netherlands, Belgium, Great Britain, Spain, India, South Korea, Vietnam, Taiwan, Colombia or Brazil
Specifications
  • Compatible WithClimMate Portable Water Activity (aW) Meter 37910-38
  • DescriptionClimMate nSens-HT-ENS Replacement Humidity and Temperature Probe for ClimMate Portable Water Activity Meter