Myron L EP11/PH Analog pH/Conductivity Meter

Representative image only.

Myron L EP11/PH Analog pH/Conductivity Meter

Myron L | Mfr: EP11/PH| Item: 0148962

Maintenance-free built-in electrodes—no replatinizing required

$1,667.73CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,667.73CAD / Each

Buy It Together

Current Item

Myron L EP11/PH Analog pH/Conductivity Meter
Myron L EP11/PH Analog pH/Conductivity Meter
0148962

$1,667.73CAD / Each
+ =
Total Price 1941.41 ADD 2 ITEMS TO CART
Product Description

Includes

One 9 V battery.

Key Features

  • Automatic temperature compensation
  • 2 year warranty

More About this Item

Get accurate, on-the-spot measurements of pH and conductivity with this compact meter. Meter features analog display of conductivity measurements from 0 to 10,000 µS in four ranges. Meter also includes a pH sensor that measures pH from 2.0 to 12.0 pH.

Analyze your samples on-site with these portable meters. All feature automatic temperature compensation and built-in cell cup. Cup contains self-maintaining electrodes that never need replatinizing. Optional conductivity range extender (sold separately) expands range by a factor of 10. Minimum sample volume is 25 mL.

The optional calibration kit 01489-51 includes a large foam-lined carrying case; 2-oz bottles of pH 4, 7 and 10 buffers; and a 7000 µS conductivity standard.

Two year warranty
Specifications
  • Meter TypePortable
  • Display typeAnalog
  • Min pH (pH)2
  • Max pH (pH)12
  • pH Resolution0.1
  • Min Conductivity0 to 10,000 µS
  • Conductivity Resolution0.2 μS
  • Conductivity Accuracy±2% FS
  • CellBuilt-in; self-conditioning
  • Temperature compensationAutomatic
  • Battery1 9V (included)
  • Battery life (hr)17520
  • Width (cm)10.16
  • Height (cm)11.43
  • Width (in)4
  • Length (in)3 1/2
  • Height (in)4 1/2
  • DescriptionAnalog pH/Conductivity Meter
  • Warranty2 year
Reviews
Q&A