Morphix 380018-10 Gas Monitoring Badges; Mercury, 10/Box
Representative image only.
Morphix 380018-10 Gas Monitoring Badges; Mercury, 10/Box
Morphix | Mfr: 380018-10| Item: 8656327
Six color-change levels to better isolate exposure readings
$430.23CAD / Box of 1
$430.23CAD / Box of 1
Product Description
Key Features
  • Ideal for both personal and area monitoring
  • Five exposure levels covering 0.15 to 1.4 mg/m3 x hr
  • Minimum detectable concentration (in 8 hours): 0.02 mg/m3
  • Relative Humidity Range: 15 to 87%
More About this Item
Gas Monitoring Badges are lightweight colorimetric — ideal for both personal and area monitoring. Designed to accurately provide immediate results while minimizing the effects of humidity and air velocity. Each ChromAir badge shows exposure levels. The scale printed on the badges is based on exposure dose. Badges attach easily to shirt or lab coat collars — order badge clips separately
Color Comparators increase the range and resolution of your ChromAir badge (order separately). The color scales on the comparator identically match the colors formed on the badges.
Specifications
  • Gases MeasuredMercury
  • SensitivityCross Sensitivity: Strong oxidizers
  • DescriptionGas Monitoring Badges; Mercury, 10/Box