Mettler Toledo SmartCal24 Moisture Analyzer Reference Substance; 24 Tests
Representative image only.
Mettler Toledo SmartCal24 Moisture Analyzer Reference Substance; 24 Tests
Mettler Toledo | Mfr: 30005790| Item: 1133631
Quick and Easy Performance Test for Moisture Analyzers
Call For Price
Call For Price
Product Description
Includes
24 test sachets.
Key Features
  • Indicates incorrect performance immediately
  • Test temperatures include 70, 100, 130, 160°C (158, 212, 266, 320°F)
  • Includes product certificate
  • Three month warranty
More About this Item
SmartCal verifies the performance of your moisture analyzer in a 10-minute test, which is carried out in the same easy way as a regular measurement. Regular testing with SmartCal lets you provides documented evidence of the unit's performance over time. Incorrect performance is immediately indicated. The test instantly indicates adverse influences from the heating or weighing units. If the test is passed, you can trust your results.
3 Month Warranty
Specifications
  • Accessory TypeFor use with Mettler Toledo Halogen Moisture Analyzer
  • DescriptionSmartCal Moisture Analyzer Reference Substance; 24 Tests