Mettler Toledo Science Pro-ISM pH Electrode; MultiPin™

Representative image only.

Mettler Toledo Science Pro-ISM pH Electrode; MultiPin™

Mettler Toledo | Mfr: 51344072| Item: 3590043

Expertly-engineered probes to cover a wide range of applications

$1,362.89CAD / Each

$1,362.89CAD / Each

Buy It Together

Current Item

Mettler Toledo Science Pro-ISM pH Electrode; MultiPin™
Mettler Toledo Science Pro-ISM pH Electrode; MultiPin™
3590043

$1,362.89CAD / Each
+ + + + + =
Total Price 4558.66 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • ISM® - Intelligent Sensor Management⌡Cable not included⌡ARGENTHAL™ reference system

More About this Item

These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.

ISM technology stores calibration data in the sensor and automatically transfer it to the meter.⌡Select the appropriate sensor cable for your application, meter, and ATC connection—BNC, waterproof BNC, DIN, and Lemo OO cables available with banana or RCA (cinch) connectors.⌡Sleeve junction provides fast results and works best in dirty samples. The A41 glass membrane works great in high tempertures and handles harsh chemicals particularly well. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.

Specifications
  • ApplicationsHarsh chemicals and high temps
  • Electrode Body MaterialGlass
  • Reference JunctionMoveable glass sleeve
  • Reference Electrolyte3M KCl
  • Sensing Glass TypeA41 - High temperatures and harsh chemicals
  • Min pH (pH)0
  • Max pH (pH)12
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • ATC ElementNTC 30 kΩ
  • ConnectorMultipin™
  • Diameter (mm)12
  • Length (mm)170
  • DescriptionpH Electrode; MultiPin™
Reviews
Q&A