Mettler Toledo S2 Handheld pH/mV Meter Standard Kit

Representative image only.

Mettler Toledo S2 Handheld pH/mV Meter Standard Kit

Mettler Toledo | Mfr: 30207950| Item: 3590346

Get quality data with one hand

$1,274.48CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,274.48CAD / Each

Buy It Together

Current Item

Mettler Toledo S2 Handheld pH/mV Meter Standard Kit
Mettler Toledo S2 Handheld pH/mV Meter Standard Kit
3590346

$1,274.48CAD / Each
+ =
Total Price 1548.16 ADD 2 ITEMS TO CART
Product Description

Includes

InLab Expert Pro-ISM-IP67 electrode, buffer sachets (4.01, 7.00, 9.21, and 10.00), electrode clip, and 4 AA batteries.

Key Features

  • User-friendly interface and single-handed T-pad operation provide fast navigation
  • Rugged IP67-rated housing protects meter in harsh environments
  • Compatible with ISM sensors—no need to recalibrate with every electrode change
  • 1 year warranty

More About this Item

These handheld pH meters are perfect for fieldwork and other mobile applications. Robust construction provides durability, while the intuitive menu and simplified button configuration let you take readings quickly.

The InLab ISM (Intelligent Sensor Management) series of electrodes stores its ID number, serial number, and calibration data. When the electrode is connected to the meter it communicates this information, so there is no need to select electrode type when switching or to recalibrate every time you power on the meter.

Choose the meter alone if you already have ISM electrodes, or choose a bundled kit suitable for your application.

Supplied electrode is general purpose and works well with emulsions and suspensions.

1 Year Warranty
Specifications
  • Meter TypePortable
  • Display typeSegmented LCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.01/0.1 pH
  • pH Accuracy±0.01 pH
  • Buffer RecognitionFour preset, one custom
  • CalibrationUp to 5 points
  • Min mV (mV)-19999
  • Max mV (mV)1999
  • mV Resolution1 mV
  • mV Accuracy±1 mV
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.5°C
  • Temperature compensationAutomatic or manual from -5.0 to 105.0°C
  • Data Logging (points)200
  • WaterproofYes
  • Battery4 AA (included)
  • Battery life (hr)400
  • Length (cm)22.2
  • Width (cm)7
  • Height (cm)3.5
  • Width (in)2 3/4
  • Length (in)8 3/4
  • Height (in)1 3/8
  • DescriptionHandheld pH/mV Meter Standard Kit
  • Warranty1 year
Reviews
Q&A