Mettler Toledo RS-P26 GXP-Compliant Compact Printer with RS-232C Interface
Representative image only.
Mettler Toledo RS-P26 GXP-Compliant Compact Printer with RS-232C Interface
Mettler Toledo | Mfr: 11124323| Item: 1133656
Print out results—remove human recording errors
Call For Price
Call For Price
Buy It Together
Current Item
Mettler Toledo RS-P26 GXP-Compliant Compact Printer with RS-232C Interface
Mettler Toledo RS-P26 GXP-Compliant Compact Printer with RS-232C Interface
1133656
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Optimize performance of your Mettler Toledo balances
  • Printer speed 2.3 lines per second
  • 24 characters per line
  • Uses standard paper
  • 3 month warranty
More About this Item
Parts and accessories needed to keep your balance operating and at its best.
3 Month Warranty
Specifications
  • USB InterfaceNo
  • RS-232 InterfaceYes
  • DescriptionRS-P26 GXP-Compliant Compact Printer with RS-232C Interface