Mettler Toledo Reusable Steel Weighing Pans, 6 mm Height; Set of 3
Representative image only.
Mettler Toledo Reusable Steel Weighing Pans, 6 mm Height; Set of 3
Mettler Toledo | Mfr: 214462| Item: 1133492
Keep your Halogen Moisture Analyzer running smoothly
$332.99CAD / Each
$332.99CAD / Each
Product Description
Key Features
  • For use with Mettler Toledo Halogen Moisture Analyzer
  • Dishwasher proof
  • Easy to wash and reuse
More About this Item
Choose from quality replacement parts and accessories to keep your Halogen Moisture Analyzer up and running.
These sample pans are the only pans officially supported by Mettler Toledo moisture analyzers for optimal use and guaranteed performance. They work well with Mettler Toledo moisture analyzer models HX204, HS153, HC103, HE73 and HE53.
Specifications
  • Accessory TypeFor use with Mettler Toledo Halogen Moisture Analyzer
  • Height (InchShort)1/4
  • DescriptionReusable Steel Weighing Pans, 6 mm Height; Set of 3