Mettler Toledo Reach 225 pH Electrode; S7
Representative image only.
Mettler Toledo Reach 225 pH Electrode; S7
Mettler Toledo | Mfr: 30244733| Item: 3590070
Expertly-engineered probes to cover a wide range of applications
$957.70CAD / Each
$957.70CAD / Each
Buy It Together
Current Item
Mettler Toledo Reach 225 pH Electrode; S7
Mettler Toledo Reach 225 pH Electrode; S7
3590070
$957.70CAD / Each
+ + + + + =
Total Price 4153.47 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Cable not included⌡HA glass membrane for high pH and temperature applications
More About this Item
These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.
Select the appropriate sensor cable for your application and meter—BNC, waterproof BNC, DIN, Lemo OO, and wire-only cables available. High alkali glass membrane provides low-alkali error—ideal for high pH and high temperature applications.
Specifications
  • ApplicationsSamples in deep containers
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference Electrolyte3M KCl
  • Sensing Glass TypeHA - High alkalifor high pH and high temp applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ATC Element
  • ConnectorS7
  • Diameter (mm)12
  • Length (mm)225
  • DescriptionpH Electrode; S7