Mettler Toledo IPac On-Site Service with ACC for Excellence XPR Precision Toploading Balances
Representative image only.
Mettler Toledo IPac On-Site Service with ACC for Excellence XPR Precision Toploading Balances
Mettler Toledo | Mfr: B99484110108| Item: 0262990
Maximize your productivity and ensure compliance
Call For Price
Call For Price
Product Description
Key Features
  • ISO 17025 Accredited Calibration Services
  • Receive installation/qualification documentation (IQ/OQ)
  • Professionally executed installation
More About this Item
IPac Standard Qualification provides professional installation by a factory-trained technician, along with installation/qualification documentation (IQ/OQ). A professionally executed installation and qualification is essential in achieving the best results from your balance. These services ensure you peace of mind that your balance performs perfectly from the start.
Specifications
  • Compatible WithExcellence XPR Precision Toploading Balances
  • DescriptionIPac On-Site Service with ACC for Excellence XPR Precision Toploading Balances